missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dithia-1,9-nonanediol 92.0+%, TCI America™

Catalog No. D28455G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28455G 5 g
D284525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28455G Supplier TCI America Supplier No. D28455G
Only null left

Chemical Identifiers

CAS 16260-48-3
Molecular Formula C7H16O2S2
Molecular Weight (g/mol) 196.32
MDL Number MFCD01098524
InChI Key JIJWGPUMOGBVMQ-UHFFFAOYSA-N
PubChem CID 373735
IUPAC Name 2-({3-[(2-hydroxyethyl)sulfanyl]propyl}sulfanyl)ethan-1-ol
SMILES OCCSCCCSCCO

Specifications

Color Yellow
Quantity 5 g
Synonym 1,3-Bis(2-hydroxyethylthio)propane, 2,2′C-(Trimethylenedithio)diethanol
Formula Weight 196.32
Percent Purity ≥92.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,7-Dithia-1,9-nonanediol