missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dimethyl-7-hydroxyoctanal, 97%

Catalog No. ac259680250
Change view
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC259680250 25 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC259680250 Supplier Thermo Scientific Chemicals Supplier No. 259680250
Only null left

CAS: 107-75-5 | C10H20O2 | 172.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 107-75-5
Molecular Formula C10H20O2
Molecular Weight (g/mol) 172.26
MDL Number MFCD00014681
InChI Key WPFVBOQKRVRMJB-UHFFFAOYSA-N
Synonym hydroxycitronellal, 7-hydroxycitronellal, citronellal hydrate, 3,7-dimethyl-7-hydroxyoctanal, phixia, cyclalia, cyclosia, laurine, fixol, lilyl aldehyde
PubChem CID 7888
ChEBI CHEBI:53459
IUPAC Name 7-hydroxy-3,7-dimethyloctanal
SMILES CC(CCCC(C)(C)O)CC=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.9200g/mL
Boiling Point 241°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4460 to 1.452
Quantity 25 mL
Specific Gravity 0.92
Formula Weight 172.26
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3, 7-Dimethyl-7-hydroxyoctanal, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,ha
EINECSNumber 203-518-7

RUO – Research Use Only