accessibility menu, dialog, popup

UserName

CAS RN 107-75-5

CH3H3CCH3HOO

CAS RN 107-75-5

IUPAC Name: 7-hydroxy-3,7-dimethyloctanal
Synonyms: hydroxycitronellal citronellal hydrate
Molecular Weight (g/mol): 172.27
Molecular Formula: C10H20O2
InChi Key: WPFVBOQKRVRMJB-UHFFFAOYNA-N
SMILES: CC(CCCC(C)(C)O)CC=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results