missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dimethyl-2,6-octadienyl Butyrate 95.0+%, TCI America™

Catalog No. D479625ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D479625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D479625ML Supplier TCI America Supplier No. D479625ML
Only null left

Chemical Identifiers

CAS 2173-82-2
Molecular Formula C14H24O2
Molecular Weight (g/mol) 224.34
MDL Number MFCD00036538
InChI Key ZSBOMYJPSRFZAL-UHFFFAOYSA-N
Synonym Butyric Acid 3,7-Dimethyl-2,6-octadienyl Ester
PubChem CID 7796
IUPAC Name 3,7-dimethylocta-2,6-dien-1-yl butanoate
SMILES CCCC(=O)OCC=C(C)CCC=C(C)C

Specifications

Color Colorless
Quantity 25 mL
Formula Weight 224.34
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,7-Dimethyl-2,6-octadienyl Butyrate
EINECSNumber (2)-0763
TSCA No