accessibility menu, dialog, popup

UserName

CAS RN 2173-82-2

H3COOCH3CH3CH3(E/Z)

CAS RN 2173-82-2

IUPAC Name: 3,7-dimethylocta-2,6-dien-1-yl butanoate
Synonyms: 3,7-dimethylocta-2,6-dien-1-yl butanoate
Molecular Weight (g/mol): 224.34
Molecular Formula: C14H24O2
InChi Key: ZSBOMYJPSRFZAL-UHFFFAOYSA-N
SMILES: CCCC(=O)OCC=C(C)CCC=C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results