missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,7-Dimethyl-1-octanol 98.0+%, TCI America™
$81.64 - $358.84
Chemical Identifiers
| CAS | 106-21-8 |
|---|---|
| Molecular Formula | C10H22O |
| Molecular Weight (g/mol) | 158.285 |
| MDL Number | MFCD00002936 |
| InChI Key | PRNCMAKCNVRZFX-UHFFFAOYSA-N |
| Synonym | 3,7-dimethyl-1-octanol, tetrahydrogeraniol, dimethyloctanol, dihydrocitronellol, pelargol, perhydrogeraniol, geraniol tetrahydride, citronellol, dihydro, 1-octanol, 3,7-dimethyl, geraniol, perhydro |
| PubChem CID | 7792 |
| IUPAC Name | 3,7-dimethyloctan-1-ol |
| SMILES | CC(C)CCCC(C)CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
D144225ML
|
N/A | 25 mL |
Each for $81.64
|
|
|||||
|
D1442500ML
|
N/A | 500 mL |
Each for $358.84
|
|
|||||
Chemical Identifiers
| C10H22O | |
| MFCD00002936 | |
| 3,7-dimethyl-1-octanol, tetrahydrogeraniol, dimethyloctanol, dihydrocitronellol, pelargol, perhydrogeraniol, geraniol tetrahydride, citronellol, dihydro, 1-octanol, 3,7-dimethyl, geraniol, perhydro | |
| 3,7-dimethyloctan-1-ol |
Specifications
| 106-21-8 | |
| 118°C | |
| MFCD00002936 | |
| PRNCMAKCNVRZFX-UHFFFAOYSA-N | |
| 3,7-dimethyloctan-1-ol | |
| 7792 | |
| ≥98.0% (GC) | |
| 3,7-Dimethyl-1-octanol |
| Colorless | |
| C10H22O | |
| 3,7-dimethyl-1-octanol, tetrahydrogeraniol, dimethyloctanol, dihydrocitronellol, pelargol, perhydrogeraniol, geraniol tetrahydride, citronellol, dihydro, 1-octanol, 3,7-dimethyl, geraniol, perhydro | |
| CC(C)CCCC(C)CCO | |
| 158.285 | |
| 158.29 | |
| Liquid |
Safety and Handling
EINECSNumber : (2)-0217
RTECSNumber : RH0900000
TSCA : Yes