missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dihydroxy-2-naphthoic Acid 90.0+%, TCI America™

Catalog No. D25165G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D25165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D25165G Supplier TCI America Supplier No. D25165G
Only null left

Specifications

CAS 83511-07-3
Molecular Formula C11H8O4
Quantity 5 g
MDL Number MFCD00010253
Synonym 3,7-Dihydroxy-2-naphthalenecarboxylic Acid
InChI Key QMWOUSYSNFCKAZ-UHFFFAOYSA-N
SMILES C1=CC(=CC2=CC(=C(C=C21)O)C(=O)O)O
IUPAC Name 3,7-dihydroxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol) 204.181
PubChem CID 736299
Formula Weight 204.18
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3,7-Dihydroxy-2-naphthoic Acid
Show More Show Less