missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Diphenylcarbazole 99.0+%, TCI America™
$76.43 - $130.48
Chemical Identifiers
| CAS | 56525-79-2 |
|---|---|
| Molecular Formula | C24H17N |
| Molecular Weight (g/mol) | 319.41 |
| MDL Number | MFCD00222619 |
| InChI Key | PCMKGEAHIZDRFL-UHFFFAOYSA-N |
| PubChem CID | 3123295 |
| IUPAC Name | 3,6-diphenyl-9H-carbazole |
| SMILES | N1C2=C(C=C(C=C2)C2=CC=CC=C2)C2=C1C=CC(=C2)C1=CC=CC=C1 |
Chemical Identifiers
| 56525-79-2 | |
| 319.41 | |
| PCMKGEAHIZDRFL-UHFFFAOYSA-N | |
| 3,6-diphenyl-9H-carbazole |
| C24H17N | |
| MFCD00222619 | |
| 3123295 | |
| N1C2=C(C=C(C=C2)C2=CC=CC=C2)C2=C1C=CC(=C2)C1=CC=CC=C1 |
Specifications
| 56525-79-2 | |
| White | |
| MFCD00222619 | |
| PCMKGEAHIZDRFL-UHFFFAOYSA-N | |
| 3,6-diphenyl-9H-carbazole | |
| 3123295 | |
| ≥99.0% (GC) | |
| 3,6-Diphenylcarbazole |
| 184°C | |
| C24H17N | |
| 1 g | |
| N1C2=C(C=C(C=C2)C2=CC=CC=C2)C2=C1C=CC(=C2)C1=CC=CC=C1 | |
| 319.41 | |
| 319.41 | |
| Crystalline Powder |
Safety and Handling
TSCA : No