accessibility menu, dialog, popup

UserName

HN

CAS RN 56525-79-2

IUPAC Name: 3,6-diphenyl-9H-carbazole
Synonyms: 3,6-diphenyl-9H-carbazole
Molecular Weight (g/mol): 319.41
Molecular Formula: C24H17N
InChi Key: PCMKGEAHIZDRFL-UHFFFAOYSA-N
SMILES: N1C2=C(C=C(C=C2)C2=CC=CC=C2)C2=C1C=CC(=C2)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results