missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 98.0+%, TCI America™
Supplier: TCI America D37435G
Spécifications
| 3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione | |
| 360°C | |
| MFCD11870113 | |
| Pigment Red 255 | |
| O=C1NC(=C2C(=O)NC(=C12)C1=CC=CC=C1)C1=CC=CC=C1 | |
| 288.31 | |
| 288.31 | |
| Crystalline Powder |
| 54660-00-3 | |
| C18H12N2O2 | |
| 5 g | |
| UWHBZLMSPZBZHY-UHFFFAOYSA-N | |
| 3,6-diphenyl-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione | |
| 5378296 | |
| ≥98.0% (N) |
Identifiants chimiques
| 54660-00-3 | |
| 288.31 | |
| UWHBZLMSPZBZHY-UHFFFAOYSA-N | |
| 5378296 | |
| O=C1NC(=C2C(=O)NC(=C12)C1=CC=CC=C1)C1=CC=CC=C1 |
| C18H12N2O2 | |
| MFCD11870113 | |
| Pigment Red 255 | |
| 3,6-diphenyl-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione |
Sécurité et manipulation
missing translation for 'einecsNumber' : (5)-5897
missing translation for 'tsca' : Yes