missing translation for 'onlineSavingsMsg'
Learn More

3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione 98.0+%, TCI America™

Catalog No. p-7127889
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D37435G 5 g
D374325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D37435G Supplier TCI America Supplier No. D37435G
Only null left

Chemical Identifiers

CAS 54660-00-3
Molecular Formula C18H12N2O2
Molecular Weight (g/mol) 288.31
MDL Number MFCD11870113
InChI Key UWHBZLMSPZBZHY-UHFFFAOYSA-N
Synonym Pigment Red 255
PubChem CID 5378296
IUPAC Name 3,6-diphenyl-1H,2H,4H,5H-pyrrolo[3,4-c]pyrrole-1,4-dione
SMILES O=C1NC(=C2C(=O)NC(=C12)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 360°C
Quantity 5 g
Formula Weight 288.31
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Diphenyl-2,5-dihydropyrrolo[3,4-c]pyrrole-1,4-dione
EINECSNumber (5)-5897
TSCA Yes