missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dimethylphenanthrene 97.0+%, TCI America™

Catalog No. D1235100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1235100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1235100MG Supplier TCI America Supplier No. D1235100MG
Only null left

Chemical Identifiers

CAS 1576-67-6
Molecular Formula C16H14
Molecular Weight (g/mol) 206.29
MDL Number MFCD00001170
InChI Key OMIBPZBOAJFEJS-UHFFFAOYSA-N
Synonym phenanthrene, 3,6-dimethyl, unii-25y8j5a29m, ccris 5483, phenanthrene,6-dimethyl, 3,6-dimethylphenantracene, acmc-1bx3v, 3,6-dimethyl phenanthrene, 3,6-dimethyl-phenanthrene, phenanthrene, 3,6-dimethyl-8ci, phenanthrene, 3,6-dimethyl-8ci 9ci
PubChem CID 15304
IUPAC Name 3,6-dimethylphenanthrene
SMILES CC1=CC=C2C=CC3=CC=C(C)C=C3C2=C1

Specifications

Melting Point 141°C
Color White-Yellow
Quantity 100 mg
Formula Weight 206.29
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dimethylphenanthrene