accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 1576-67-6

IUPAC Name: 3,6-dimethylphenanthrene
Synonyms: 3,6-dimethylphenanthrene
Molecular Weight (g/mol): 206.29
Molecular Formula: C16H14
InChi Key: OMIBPZBOAJFEJS-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC3=CC=C(C)C=C3C2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results