missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dimethyl-4-octyne-3,6-diol 98.0+%, TCI America™

Catalog No. D1168500G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1168500G 500 g
D116825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D1168500G Supplier TCI America Supplier No. D1168500G

Chemical Identifiers

CAS 78-66-0
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.252
MDL Number MFCD00004480
InChI Key NUYADIDKTLPDGG-UHFFFAOYSA-N
Synonym 3,6-Dihydroxy-3,6-dimethyl-4-octyne
PubChem CID 6546
IUPAC Name 3,6-dimethyloct-4-yne-3,6-diol
SMILES CCC(C)(C#CC(C)(CC)O)O

Specifications

Melting Point 54°C
Color White
Boiling Point 135°C
Quantity 500 g
Formula Weight 170.25
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dimethyl-4-octyne-3,6-diol
EINECSNumber (2)-0269
RTECSNumber RI2650000
TSCA Yes