missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dimethoxy-9H-carbazole 98.0+%, TCI America™
Supplier: TCI America D4833200MG
Specifications
| 3,6-Dimethoxy-9H-carbazole | |
| 120°C | |
| C14H13NO2 | |
| 200 mg | |
| COC1=CC2=C(NC3=C2C=C(OC)C=C3)C=C1 | |
| 227.26 | |
| 227.26 | |
| Crystalline Powder |
| 57103-01-2 | |
| White | |
| MFCD00626046 | |
| YQKMWXHJSIEAEX-UHFFFAOYSA-N | |
| 3,6-dimethoxy-9H-carbazole | |
| 644464 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 57103-01-2 | |
| 227.26 | |
| YQKMWXHJSIEAEX-UHFFFAOYSA-N | |
| 3,6-dimethoxy-9H-carbazole |
| C14H13NO2 | |
| MFCD00626046 | |
| 644464 | |
| COC1=CC2=C(NC3=C2C=C(OC)C=C3)C=C1 |
Safety and Handling
TSCA : No