missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dimethoxy-9H-carbazole 98.0+%, TCI America™

Catalog No. p-7128492
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4833200MG 200 mg
D48331G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4833200MG Supplier TCI America Supplier No. D4833200MG
Only null left

Chemical Identifiers

CAS 57103-01-2
Molecular Formula C14H13NO2
Molecular Weight (g/mol) 227.26
MDL Number MFCD00626046
InChI Key YQKMWXHJSIEAEX-UHFFFAOYSA-N
PubChem CID 644464
IUPAC Name 3,6-dimethoxy-9H-carbazole
SMILES COC1=CC2=C(NC3=C2C=C(OC)C=C3)C=C1

Specifications

Melting Point 120°C
Color White
Quantity 200 mg
Formula Weight 227.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dimethoxy-9H-carbazole