missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Diiodo-9-phenylcarbazole 98.0+%, TCI America™
$72.70 - $374.68
Chemical Identifiers
| CAS | 57103-21-6 |
|---|---|
| Molecular Formula | C18H11I2N |
| Molecular Weight (g/mol) | 495.102 |
| MDL Number | MFCD11521285 |
| InChI Key | AWGAUYXFWGUFNE-UHFFFAOYSA-N |
| PubChem CID | 13407347 |
| IUPAC Name | 3,6-diiodo-9-phenylcarbazole |
| SMILES | C1=CC=C(C=C1)N2C3=C(C=C(C=C3)I)C4=C2C=CC(=C4)I |
Chemical Identifiers
| 57103-21-6 | |
| 495.102 | |
| AWGAUYXFWGUFNE-UHFFFAOYSA-N | |
| 3,6-diiodo-9-phenylcarbazole |
| C18H11I2N | |
| MFCD11521285 | |
| 13407347 | |
| C1=CC=C(C=C1)N2C3=C(C=C(C=C3)I)C4=C2C=CC(=C4)I |
Specifications
| 57103-21-6 | |
| C18H11I2N | |
| 1 g | |
| C1=CC=C(C=C1)N2C3=C(C=C(C=C3)I)C4=C2C=CC(=C4)I | |
| 495.102 | |
| 495.10 | |
| Crystalline Powder |
| 183°C | |
| MFCD11521285 | |
| AWGAUYXFWGUFNE-UHFFFAOYSA-N | |
| 3,6-diiodo-9-phenylcarbazole | |
| 13407347 | |
| ≥98.0% (GC) | |
| 3,6-Diiodo-9-phenylcarbazole |
Safety and Handling
TSCA : No