accessibility menu, dialog, popup

UserName

NII

CAS RN 57103-21-6

IUPAC Name: 3,6-diiodo-9-phenyl-9H-carbazole
Synonyms: 3,6-diiodo-9-phenylcarbazole
Molecular Weight (g/mol): 495.1
Molecular Formula: C18H11I2N
InChi Key: AWGAUYXFWGUFNE-UHFFFAOYSA-N
SMILES: IC1=CC2=C(C=C1)N(C1=C2C=C(I)C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results