Learn More
3,6-Difluorophthalic acid, 98%, Thermo Scientific Chemicals
$330.11 - $330.11
Chemical Identifiers
| CAS | 651-97-8 |
|---|---|
| Molecular Formula | C8H4F2O4 |
| Molecular Weight (g/mol) | 202.113 |
| MDL Number | MFCD03840499 |
| InChI Key | VFLMWMTWWZGXGA-UHFFFAOYSA-N |
| Synonym | 3,6-difluorobenzene-1,2-dicarboxylic acid, 1,4-difluorophthalic acid, 3,6-difluorophthalicacid, pubchem1945, acmc-1ays1, 3,6-difluoro-phthalic acid, ksc352e6d, 3,5-difluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 3,6-difluoro |
| PubChem CID | 643385 |
| IUPAC Name | 3,6-difluorophthalic acid |
| SMILES | C1=CC(=C(C(=C1F)C(=O)O)C(=O)O)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2609603
|
Thermo Scientific Chemicals
H2609603 |
1 g |
Each for $330.11
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 651-97-8 | |
| 202.113 | |
| VFLMWMTWWZGXGA-UHFFFAOYSA-N | |
| 643385 | |
| C1=CC(=C(C(=C1F)C(=O)O)C(=O)O)F |
| C8H4F2O4 | |
| MFCD03840499 | |
| 3,6-difluorobenzene-1,2-dicarboxylic acid, 1,4-difluorophthalic acid, 3,6-difluorophthalicacid, pubchem1945, acmc-1ays1, 3,6-difluoro-phthalic acid, ksc352e6d, 3,5-difluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 3,6-difluoro | |
| 3,6-difluorophthalic acid |
Specifications
| 651-97-8 | |
| C8H4F2O4 | |
| 1 g | |
| VFLMWMTWWZGXGA-UHFFFAOYSA-N | |
| 3,6-difluorophthalic acid | |
| 643385 | |
| 98% |
| ∼184°C (decomposition) | |
| MFCD03840499 | |
| 3,6-difluorobenzene-1,2-dicarboxylic acid, 1,4-difluorophthalic acid, 3,6-difluorophthalicacid, pubchem1945, acmc-1ays1, 3,6-difluoro-phthalic acid, ksc352e6d, 3,5-difluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 3,6-difluoro | |
| C1=CC(=C(C(=C1F)C(=O)O)C(=O)O)F | |
| 202.113 | |
| 202.11 | |
| 3,6-Difluorophthalic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only