missing translation for 'onlineSavingsMsg'
Learn More

3,6-Difluorophthalic acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH2609603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2609603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH2609603 Supplier Thermo Scientific Chemicals Supplier No. H2609603

CAS: 651-97-8 | Molecular Formula: C8H4F2O4 | Molecular Weight: 202.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 651-97-8
Molecular Formula C8H4F2O4
Molecular Weight (g/mol) 202.113
MDL Number MFCD03840499
InChI Key VFLMWMTWWZGXGA-UHFFFAOYSA-N
Synonym 3,6-difluorobenzene-1,2-dicarboxylic acid, 1,4-difluorophthalic acid, 3,6-difluorophthalicacid, pubchem1945, acmc-1ays1, 3,6-difluoro-phthalic acid, ksc352e6d, 3,5-difluoro-1,2-benzenedicarboxylic acid, 1,2-benzenedicarboxylicacid, 3,6-difluoro
PubChem CID 643385
IUPAC Name 3,6-difluorophthalic acid
SMILES C1=CC(=C(C(=C1F)C(=O)O)C(=O)O)F

Specifications

Melting Point ∼184°C (decomposition)
Quantity 1 g
Formula Weight 202.11
Percent Purity 98%
Chemical Name or Material 3,6-Difluorophthalic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only