missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dibromopyridazine 98.0+%, TCI America™
$59.65 - $208.78
Chemical Identifiers
| CAS | 17973-86-3 |
|---|---|
| Molecular Formula | C4H2Br2N2 |
| Molecular Weight (g/mol) | 237.882 |
| MDL Number | MFCD00233947 |
| InChI Key | VQAFMTSSCUETHA-UHFFFAOYSA-N |
| PubChem CID | 248852 |
| IUPAC Name | 3,6-dibromopyridazine |
| SMILES | C1=CC(=NN=C1Br)Br |
Chemical Identifiers
| 17973-86-3 | |
| 237.882 | |
| VQAFMTSSCUETHA-UHFFFAOYSA-N | |
| 3,6-dibromopyridazine |
| C4H2Br2N2 | |
| MFCD00233947 | |
| 248852 | |
| C1=CC(=NN=C1Br)Br |
Specifications
| 17973-86-3 | |
| White-Yellow | |
| MFCD00233947 | |
| 3,6-dibromopyridazide, pyridazine, 3,6-dibromo, 3,6-dibromo pyridazine, pubchem12385, 3,6-dibromo-pyridazine, pyridazine,3,6-dibromo, 3,6-dibromo-1,2-diazine, 3,6-dibromopyridazine, synquest 3h32-9-t3 | |
| C1=CC(=NN=C1Br)Br | |
| 237.882 | |
| 237.88 | |
| Crystalline Powder |
| 119°C | |
| C4H2Br2N2 | |
| 1 g | |
| VQAFMTSSCUETHA-UHFFFAOYSA-N | |
| 3,6-dibromopyridazine | |
| 248852 | |
| ≥98.0% (GC) | |
| 3,6-Dibromopyridazine |
Safety and Handling
TSCA : No