accessibility menu, dialog, popup

UserName

BrBrNN

CAS RN 17973-86-3

IUPAC Name: 3,6-dibromopyridazine
Synonyms: 3,6-dibromopyridazine
Molecular Weight (g/mol): 237.88
Molecular Formula: C4H2Br2N2
InChi Key: VQAFMTSSCUETHA-UHFFFAOYSA-N
SMILES: BrC1=CC=C(Br)N=N1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)