missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromopyridazine 98.0+%, TCI America™

Catalog No. D54661G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54661G 1 g
D54665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54661G Supplier TCI America Supplier No. D54661G

Chemical Identifiers

CAS 17973-86-3
Molecular Formula C4H2Br2N2
Molecular Weight (g/mol) 237.882
MDL Number MFCD00233947
InChI Key VQAFMTSSCUETHA-UHFFFAOYSA-N
PubChem CID 248852
IUPAC Name 3,6-dibromopyridazine
SMILES C1=CC(=NN=C1Br)Br

Specifications

Melting Point 119°C
Color White-Yellow
Quantity 1 g
Synonym 3,6-dibromopyridazide, pyridazine, 3,6-dibromo, 3,6-dibromo pyridazine, pubchem12385, 3,6-dibromo-pyridazine, pyridazine,3,6-dibromo, 3,6-dibromo-1,2-diazine, 3,6-dibromopyridazine, synquest 3h32-9-t3
Formula Weight 237.88
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromopyridazine