missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromophthalic Anhydride 98.0+%, TCI America™

Catalog No. p-7128424
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4724200MG 200 mg
D47241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D4724200MG Supplier TCI America Supplier No. D4724200MG

Chemical Identifiers

CAS 25834-16-6
Molecular Formula C8H2Br2O3
Molecular Weight (g/mol) 305.91
MDL Number MFCD01117424
InChI Key OUAOHMOFJGFOSR-UHFFFAOYSA-N
PubChem CID 54169537
IUPAC Name 4,7-dibromo-1,3-dihydro-2-benzofuran-1,3-dione
SMILES BrC1=C2C(=O)OC(=O)C2=C(Br)C=C1

Specifications

Melting Point 209°C
Color White
Quantity 200 mg
Formula Weight 305.91
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromophthalic Anhydride
EINECSNumber (3)-0062
TSCA No