accessibility menu, dialog, popup

UserName

BrOOOBr

CAS RN 25834-16-6

IUPAC Name: 4,7-dibromo-1,3-dihydro-2-benzofuran-1,3-dione
Synonyms: 4,7-dibromo-2-benzofuran-1,3-dione
Molecular Weight (g/mol): 305.91
Molecular Formula: C8H2Br2O3
InChi Key: OUAOHMOFJGFOSR-UHFFFAOYSA-N
SMILES: BrC1=C2C(=O)OC(=O)C2=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results