missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromocarbazole 98.0+%, TCI America™

Catalog No. D29831G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29831G 1 g
D29835G 5 g
D298325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D29831G Supplier TCI America Supplier No. D29831G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6825-20-3
Molecular Formula C12H7Br2N
Molecular Weight (g/mol) 325.003
InChI Key FIHILUSWISKVSR-UHFFFAOYSA-N
Synonym 3,6-dibromocarbazole, 9h-carbazole, 3,6-dibromo, carbazole, 3,6-dibromo, pubchem9951, 3,6-dibromo-carbazole, acmc-209o2m, 3,6,-dibromo-9h-carbazole, 3,6-dibromo-9-h-carbazole, ksc490o8t
PubChem CID 274874
IUPAC Name 3,6-dibromo-9H-carbazole
SMILES C1=CC2=C(C=C1Br)C3=C(N2)C=CC(=C3)Br

Specifications

Melting Point 212°C
Quantity 1 g
Formula Weight 325.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromocarbazole