missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-9-phenylcarbazole 98.0+%, TCI America™

Catalog No. p-7127590
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D29811G 1 g
D29815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D29811G Supplier TCI America Supplier No. D29811G

Chemical Identifiers

CAS 57103-20-5
Molecular Formula C18H11Br2N
Molecular Weight (g/mol) 401.101
InChI Key JBWRZTKHMKVFMQ-UHFFFAOYSA-N
Synonym 3,6-dibromo-9-phenyl-9h-carbazole, 9h-carbazole, 3,6-dibromo-9-phenyl, 3,6-dibromo-9-phenyl-carbazole, ksc268e0n, 3,6-dibromo-9-phenylarbazole, n-phenyl-3,6-dibromocarbazole, acmc-1b118, 3,6-dibromo-9-phenylcarbazole
PubChem CID 11025791
IUPAC Name 3,6-dibromo-9-phenylcarbazole
SMILES C1=CC=C(C=C1)N2C3=C(C=C(C=C3)Br)C4=C2C=CC(=C4)Br

Specifications

Quantity 1 g
Formula Weight 401.10
Percent Purity ≥98.0% (GC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-phenylcarbazole