accessibility menu, dialog, popup

UserName

CAS RN 57103-20-5

IUPAC Name: 3,6-dibromo-9-phenyl-9H-carbazole
Synonyms: 3,6-dibromo-9-phenylcarbazole
Molecular Weight (g/mol): 401.1
Molecular Formula: C18H11Br2N
InChi Key: JBWRZTKHMKVFMQ-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(C=C1)N(C1=C2C=C(Br)C=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results