missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Dibromo-9-hexyl-9H-carbazole 98.0+%, TCI America™
$223.68 - $648.70
Chemical Identifiers
| CAS | 150623-72-6 |
|---|---|
| Molecular Formula | C18H19Br2N |
| Molecular Weight (g/mol) | 409.17 |
| MDL Number | MFCD23099108 |
| InChI Key | POMJRCRJZRALHK-UHFFFAOYSA-N |
| PubChem CID | 12174281 |
| IUPAC Name | 3,6-dibromo-9-hexyl-9H-carbazole |
| SMILES | CCCCCCN1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2 |
Chemical Identifiers
| 150623-72-6 | |
| 409.17 | |
| POMJRCRJZRALHK-UHFFFAOYSA-N | |
| 3,6-dibromo-9-hexyl-9H-carbazole |
| C18H19Br2N | |
| MFCD23099108 | |
| 12174281 | |
| CCCCCCN1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2 |
Specifications
| 150623-72-6 | |
| C18H19Br2N | |
| 1 g | |
| CCCCCCN1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2 | |
| 409.17 | |
| 409.17 | |
| Crystalline Powder |
| 100°C | |
| MFCD23099108 | |
| POMJRCRJZRALHK-UHFFFAOYSA-N | |
| 3,6-dibromo-9-hexyl-9H-carbazole | |
| 12174281 | |
| ≥98.0% (GC) | |
| 3,6-Dibromo-9-hexyl-9H-carbazole |
Safety and Handling
TSCA : No