accessibility menu, dialog, popup

UserName

H3CNBrBr

CAS RN 150623-72-6

IUPAC Name: 3,6-dibromo-9-hexyl-9H-carbazole
Synonyms: 3,6-dibromo-9-hexylcarbazole
Molecular Weight (g/mol): 409.17
Molecular Formula: C18H19Br2N
InChi Key: POMJRCRJZRALHK-UHFFFAOYSA-N
SMILES: CCCCCCN1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results