missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-9-hexyl-9H-carbazole 98.0+%, TCI America™

Catalog No. D54105G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54105G 5 g
D54101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54105G Supplier TCI America Supplier No. D54105G
Only null left

Chemical Identifiers

CAS 150623-72-6
Molecular Formula C18H19Br2N
Molecular Weight (g/mol) 409.17
MDL Number MFCD23099108
InChI Key POMJRCRJZRALHK-UHFFFAOYSA-N
PubChem CID 12174281
IUPAC Name 3,6-dibromo-9-hexyl-9H-carbazole
SMILES CCCCCCN1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2

Specifications

Melting Point 100°C
Quantity 5 g
Formula Weight 409.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-hexyl-9H-carbazole