missing translation for 'onlineSavingsMsg'
Learn More

3,6-Dibromo-2-methylpyridine 98.0+%, TCI America™

Catalog No. p-7128264
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44151G 1 g
D44155G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44151G Supplier TCI America Supplier No. D44151G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 39919-65-8 | C6H5Br2N | 250.92 g/mol

Chemical Identifiers

CAS 39919-65-8
Molecular Formula C6H5Br2N
Molecular Weight (g/mol) 250.92
MDL Number MFCD06254589
InChI Key UCHKRHGVKYVGTC-UHFFFAOYSA-N
Synonym 2,5-dibromo-6-methylpyridine, 3,6-dibromo-2-picoline, pyridine, 3,6-dibromo-2-methyl, 3,6-dibromo-2-methyl-pyridine, pubchem3580, 2,5-dibromo-6-picoline, 2,5-dibromo-6-methypyridine, ksc495i3h, 2,5-dibromo-6-methyl pyridine, 3,6-dibromo-2-methylpyridine
PubChem CID 2762946
IUPAC Name 3,6-dibromo-2-methylpyridine
SMILES CC1=C(Br)C=CC(Br)=N1

Specifications

Melting Point 38°C
Color White-Yellow
Quantity 1 g
Formula Weight 250.92
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-2-methylpyridine