accessibility menu, dialog, popup

UserName

CAS RN 39919-65-8

CH3NBrBr

CAS RN 39919-65-8

IUPAC Name: 3,6-dibromo-2-methylpyridine
Synonyms: 3,6-dibromo-2-methylpyridine
Molecular Weight (g/mol): 250.92
Molecular Formula: C6H5Br2N
InChi Key: UCHKRHGVKYVGTC-UHFFFAOYSA-N
SMILES: CC1=C(Br)C=CC(Br)=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results