missing translation for 'onlineSavingsMsg'
Learn More

3,6-Di-tert-butylfluorene, 98%

Catalog No. AAB2399603
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2399603 1 g
AAB2399606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2399603 Supplier Thermo Scientific Chemicals Supplier No. B2399603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 58775-07-8 | C21H26 | 278.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 58775-07-8
Molecular Formula C21H26
Molecular Weight (g/mol) 278.44
MDL Number MFCD02683109
InChI Key IWUHFBNEZZKUEY-UHFFFAOYSA-N
PubChem CID 11140437
SMILES CC(C)(C)C1=CC2=C(CC3=C2C=C(C=C3)C(C)(C)C)C=C1

Specifications

Melting Point 79°C to 82°C
Quantity 1 g
Synonym 3,6-di-tert-butylfluorene, 3,6-di-tert-butyl-9h-fluorene, acmc-1atta, 3,6-di-t-butylfluorene, 3,6-di-tert-butyl-fluorene, 3,6-bis tert-butyl fluorene
IUPAC Name 3,6-ditert-butyl-9H-fluorene
Formula Weight 278.44
Percent Purity 98%
Chemical Name or Material 3,6-Di-tert-butylfluorene
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only