missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Di-tert-butylcarbazole 98.0+%, TCI America™
$147.25 - $419.41
Chemical Identifiers
| CAS | 37500-95-1 |
|---|---|
| Molecular Formula | C20H25N |
| Molecular Weight (g/mol) | 279.43 |
| MDL Number | MFCD03425849 |
| InChI Key | OYFFSPILVQLRQA-UHFFFAOYSA-N |
| PubChem CID | 15469209 |
| IUPAC Name | 3,6-di-tert-butyl-9H-carbazole |
| SMILES | CC(C)(C)C1=CC2=C(NC3=C2C=C(C=C3)C(C)(C)C)C=C1 |
Chemical Identifiers
| 37500-95-1 | |
| 279.43 | |
| OYFFSPILVQLRQA-UHFFFAOYSA-N | |
| 3,6-di-tert-butyl-9H-carbazole |
| C20H25N | |
| MFCD03425849 | |
| 15469209 | |
| CC(C)(C)C1=CC2=C(NC3=C2C=C(C=C3)C(C)(C)C)C=C1 |
Specifications
| 37500-95-1 | |
| White | |
| MFCD03425849 | |
| OYFFSPILVQLRQA-UHFFFAOYSA-N | |
| 3,6-di-tert-butyl-9H-carbazole | |
| 15469209 | |
| ≥98.0% (GC) | |
| 3,6-Di-tert-butylcarbazole |
| 232°C | |
| C20H25N | |
| 1 g | |
| CC(C)(C)C1=CC2=C(NC3=C2C=C(C=C3)C(C)(C)C)C=C1 | |
| 279.43 | |
| 279.43 | |
| Crystalline Powder |
Safety and Handling
TSCA : No