missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,6-Di-tert-butylcarbazole 98.0+%, TCI America™
Supplier: TCI America D39525G
Specifications
| 3,6-Di-tert-butylcarbazole | |
| 232°C | |
| C20H25N | |
| 5 g | |
| CC(C)(C)C1=CC2=C(NC3=C2C=C(C=C3)C(C)(C)C)C=C1 | |
| 279.43 | |
| 279.43 | |
| Crystalline Powder |
| 37500-95-1 | |
| White | |
| MFCD03425849 | |
| OYFFSPILVQLRQA-UHFFFAOYSA-N | |
| 3,6-di-tert-butyl-9H-carbazole | |
| 15469209 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 37500-95-1 | |
| 279.43 | |
| OYFFSPILVQLRQA-UHFFFAOYSA-N | |
| 3,6-di-tert-butyl-9H-carbazole |
| C20H25N | |
| MFCD03425849 | |
| 15469209 | |
| CC(C)(C)C1=CC2=C(NC3=C2C=C(C=C3)C(C)(C)C)C=C1 |
Safety and Handling
TSCA : No