missing translation for 'onlineSavingsMsg'
Learn More

3,6-Bis(bromomethyl)durene 98.0+%, TCI America™

Catalog No. B21585G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B21585G Supplier TCI America Supplier No. B21585G
Only null left

Chemical Identifiers

CAS 35168-64-0
Molecular Formula C12H16Br2
Molecular Weight (g/mol) 320.07
MDL Number MFCD01114746
InChI Key OREPDRBSWICSCF-UHFFFAOYSA-N
Synonym 1,4-Bis(bromomethyl)-2,3,5,6-tetramethylbenzene
PubChem CID 2752645
IUPAC Name 1,4-bis(bromomethyl)-2,3,5,6-tetramethylbenzene
SMILES CC1=C(C)C(CBr)=C(C)C(C)=C1CBr

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 320.07
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Bis(bromomethyl)durene