missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Nitro-2-furyl)acrylic Acid 97.0+%, TCI America™

Catalog No. N05515G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N05515G 5 g
N055125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N05515G Supplier TCI America Supplier No. N05515G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 6281-23-8
Molecular Formula C7H5NO5
Molecular Weight (g/mol) 183.119
MDL Number MFCD00143078
InChI Key LWOWNIPZHGWKNR-DUXPYHPUSA-N
PubChem CID 702695
SMILES C1=C(OC(=C1)[N+](=O)[O-])C=CC(=O)O

Specifications

Quantity 5 g
Formula Weight 183.12
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 3-(5-Nitro-2-furyl)acrylic Acid
RTECSNumber LT8566000
TSCA No