accessibility menu, dialog, popup

UserName

HOOONOO(E)

CAS RN 6281-23-8

IUPAC Name: (2E)-3-(5-nitrofuran-2-yl)prop-2-enoic acid
Synonyms: (2E)-3-(5-nitrofuran-2-yl)prop-2-enoic acid
Molecular Weight (g/mol): 183.12
Molecular Formula: C7H5NO5
InChi Key: LWOWNIPZHGWKNR-DUXPYHPUSA-N
SMILES: OC(=O)\C=C\C1=CC=C(O1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results