missing translation for 'onlineSavingsMsg'
Learn More

3-(5-Methyl-1,2,4-oxadiazol-3-yl)benzenesulfonyl chloride, Tech., Thermo Scientific™

Catalog No. CC39103CB
Change view
Click to view available options
Quantity:
250mg
1g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC39103CB 250mg Amber Glass Bottle
CC39103DA 1g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC39103CB Supplier Thermo Scientific Chemicals Supplier No. CC39103CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 10185-62-3
Molecular Formula C9H7ClN2O3S
Molecular Weight (g/mol) 258.68
MDL Number MFCD06797486
InChI Key YPMVPGZXUILSPT-UHFFFAOYSA-N
Synonym 3-5-methyl-1,2,4-oxadiazol-3-yl benzenesulfonyl chloride, benzenesulfonylchloride,3-5-methyl-1,2,4-oxadiazol-3-yl, 3-5-methyl-1,2,4-oxadiazol-3-yl benzene-1-sulfonyl chloride, chloro 3-5-methyl 1,2,4-oxadiazol-3-yl phenyl sulfone, 3-5-methyl-1,2,4 oxadiazol-3-yl-benzenesulfonylchloride, 3-5-methyl-1,2,4-oxadiazol-3-yl phenylsulfonyl chloride, benzenesulfonyl chloride, 3-5-methyl-1,2,4-oxadiazol-3-yl, benzenesulfonylchloride, 3-5-methyl-1,2,4-oxadiazol-3-yl
PubChem CID 18525745
IUPAC Name 3-(5-methyl-1,2,4-oxadiazol-3-yl)benzenesulfonyl chloride
SMILES CC1=NC(=NO1)C1=CC(=CC=C1)S(Cl)(=O)=O

Specifications

CAS Min % 80.0
CAS Max % 100.0
Packaging Amber Glass Bottle
Quantity 250mg
Formula Weight 258.68
Percent Purity Tech.
Grade Technical
Chemical Name or Material 3-(5-Methyl-1, 2, 4-oxadiazol-3-yl)benzenesulfonyl chloride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.