accessibility menu, dialog, popup

UserName

CAS RN 10185-62-3

CH3NNOSOOCl

CAS RN 10185-62-3

IUPAC Name: 3-(5-methyl-1,2,4-oxadiazol-3-yl)benzene-1-sulfonyl chloride
Synonyms: 3-(5-methyl-1,2,4-oxadiazol-3-yl)benzenesulfonyl chloride
Molecular Weight (g/mol): 258.68
Molecular Formula: C9H7ClN2O3S
InChi Key: YPMVPGZXUILSPT-UHFFFAOYSA-N
SMILES: CC1=NC(=NO1)C1=CC(=CC=C1)S(Cl)(=O)=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)