missing translation for 'onlineSavingsMsg'
Learn More

3,5-Heptanedione, 99%

Catalog No. AC411260050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC411260050 5 g Glass bottle
AC411260250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC411260050 Supplier Thermo Scientific Chemicals Supplier No. 411260050

CAS: 7424-54-6 | C7H12O2 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7424-54-6
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00015186
InChI Key DGCTVLNZTFDPDJ-UHFFFAOYSA-N
Synonym 3,5-heptanedione, dipropionylmethane, 3,5-heptandione, unii-q3b558e3vy, 3,5-heptadione, 3,5-heptane dione, pubchem21961, ksc379q6f, 3,5-heptanedione, purum gc
PubChem CID 81923
IUPAC Name heptane-3,5-dione
SMILES CCC(=O)CC(=O)CC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.9450g/mL
Boiling Point 172°C to 173°C (711.0 mmHg)
Flash Point 57°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4540 to 1.4570 (20°C, 589nm)
Linear Formula CH3CH2COCH2COCH2CH3
Quantity 5 g
Specific Gravity 0.945
Formula Weight 128.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3, 5-Heptanedione
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 231-054-5
TSCA TSCA

RUO – Research Use Only