missing translation for 'onlineSavingsMsg'
Learn More

3,5-Heptanedione, 97%

CAS: 7424-54-6 | C7H12O2 | 128.171 g/mol

$119.68 - $395.42

Chemical Identifiers

CAS 7424-54-6
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.171
MDL Number MFCD00015186
InChI Key DGCTVLNZTFDPDJ-UHFFFAOYSA-N
Synonym 3,5-heptanedione, dipropionylmethane, 3,5-heptandione, unii-q3b558e3vy, 3,5-heptadione, 3,5-heptane dione, pubchem21961, ksc379q6f, 3,5-heptanedione, purum gc
PubChem CID 81923
IUPAC Name heptane-3,5-dione
SMILES CCC(=O)CC(=O)CC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1816506
View Documents
Thermo Scientific Chemicals
A1816506
5 g
Each for $119.68
Only null left
 
AAA1816514
View Documents
Thermo Scientific Chemicals
A1816514
25 g
Each for $395.42
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7424-54-6
128.171
DGCTVLNZTFDPDJ-UHFFFAOYSA-N
81923
CCC(=O)CC(=O)CC
C7H12O2
MFCD00015186
3,5-heptanedione, dipropionylmethane, 3,5-heptandione, unii-q3b558e3vy, 3,5-heptadione, 3,5-heptane dione, pubchem21961, ksc379q6f, 3,5-heptanedione, purum gc
heptane-3,5-dione

Specifications

7424-54-6
175°C to 177°C
Pleasant
1.4555
5 g
635979
DGCTVLNZTFDPDJ-UHFFFAOYSA-N
heptane-3,5-dione
81923
98%
0.8
57°C (134°F)
C7H12O2
MFCD00015186
UN1224
3,5-heptanedione, dipropionylmethane, 3,5-heptandione, unii-q3b558e3vy, 3,5-heptadione, 3,5-heptane dione, pubchem21961, ksc379q6f, 3,5-heptanedione, purum gc
CCC(=O)CC(=O)CC
128.171
128.17
3,5-Heptanedione
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

EINECSNumber : 231-054-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only