missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Diphenyl-1-(4-biphenylyl)formazan 95.0+%, TCI America™
$50.33 - $301.98
Chemical Identifiers
| CAS | 21520-85-4 |
|---|---|
| Molecular Formula | C25H20N4 |
| Molecular Weight (g/mol) | 376.463 |
| MDL Number | MFCD00059680 |
| InChI Key | LKFXJYZDDITMNN-UHFFFAOYSA-N |
| Synonym | 3,5-Diphenyl-1-(p-diphenylyl)formazan |
| PubChem CID | 53424949 |
| IUPAC Name | N'-anilino-N-(4-phenylphenyl)iminobenzenecarboximidamide |
| SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)N=NC(=NNC3=CC=CC=C3)C4=CC=CC=C4 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B0277100MG
|
TCI America
B0277100MG |
100 mg |
Each for $50.33
|
|
|||||
|
B02771G
|
TCI America
B02771G |
1 g |
Each for $301.98
|
|
|||||
Chemical Identifiers
| 21520-85-4 | |
| 376.463 | |
| LKFXJYZDDITMNN-UHFFFAOYSA-N | |
| 53424949 | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)N=NC(=NNC3=CC=CC=C3)C4=CC=CC=C4 |
| C25H20N4 | |
| MFCD00059680 | |
| 3,5-Diphenyl-1-(p-diphenylyl)formazan | |
| N'-anilino-N-(4-phenylphenyl)iminobenzenecarboximidamide |
Specifications
| 21520-85-4 | |
| Purple-Red | |
| MFCD00059680 | |
| 3,5-Diphenyl-1-(p-diphenylyl)formazan | |
| C1=CC=C(C=C1)C2=CC=C(C=C2)N=NC(=NNC3=CC=CC=C3)C4=CC=CC=C4 | |
| 376.463 | |
| 376.46 | |
| Crystalline Powder |
| 169°C | |
| C25H20N4 | |
| 100 mg | |
| LKFXJYZDDITMNN-UHFFFAOYSA-N | |
| N'-anilino-N-(4-phenylphenyl)iminobenzenecarboximidamide | |
| 53424949 | |
| ≥95.0% (HPLC) | |
| 3,5-Diphenyl-1-(4-biphenylyl)formazan |
Safety and Handling
TSCA : No