CAS RN 21520-85-4
CAS RN 21520-85-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,5-Diphenyl-1-(4-biphenylyl)formazan 95.0+%, TCI America™
CAS: 21520-85-4 Molecular Formula: C25H20N4 Molecular Weight (g/mol): 376.463 MDL Number: MFCD00059680 InChI Key: LKFXJYZDDITMNN-UHFFFAOYSA-N Synonym: 3,5-Diphenyl-1-(p-diphenylyl)formazan PubChem CID: 53424949 IUPAC Name: N'-anilino-N-(4-phenylphenyl)iminobenzenecarboximidamide SMILES: C1=CC=C(C=C1)C2=CC=C(C=C2)N=NC(=NNC3=CC=CC=C3)C4=CC=CC=C4