missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrosalicylaldehyde, 98%

Catalog No. AAB2042403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2042403 1 g
AAB2042406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2042403 Supplier Thermo Scientific Chemicals Supplier No. B2042403

CAS: 2460-59-5 | C7H3N2O6 | 211.11 g/mol

It is used to produce 3-(2-hydroxy-3,5-dinitro-phenyl)-propenal with acetaldehyde. 3,5-Dinitrosalicylaldehyde has been used in the preparation of salicyldimine ligand via Schiff base condensation with allyl-substituted aniline, 3-hydroxycoumarins2, chromogenic proteinase substrates and ruthenium(II) chiral Schiff base complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce 3-(2-hydroxy-3,5-dinitro-phenyl)-propenal with acetaldehyde. 3,5-Dinitrosalicylaldehyde has been used in the preparation of salicyldimine ligand via Schiff base condensation with allyl-substituted aniline, 3-hydroxycoumarins2, chromogenic proteinase substrates and ruthenium(II) chiral Schiff base complexes.

Solubility
Soluble in water, 0.6 g/100 mL (18°C).

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Chemical Identifiers

CAS 2460-59-5
Molecular Formula C7H3N2O6
Molecular Weight (g/mol) 211.11
MDL Number MFCD00007103
InChI Key FLJXIBHYDIMYRS-UHFFFAOYSA-M
PubChem CID 75571
IUPAC Name 2-formyl-4,6-dinitrobenzen-1-olate
SMILES [O-]C1=C(C=O)C=C(C=C1[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 71°C to 74°C
Quantity 1 g
Beilstein 1121900
Sensitivity Air Sensitive
Synonym 3,5-dinitrosalicylaldehyde, 2-hydroxy-3,5-dinitrobenzaldehyde, 3,5-dinitro-2-hydroxybenzaldehyde, benzaldehyde, 2-hydroxy-3,5-dinitro, acmc-20anby, 2-hydroxy-3,5-dinitro-benzaldehyde, 2-hydroxy-3,5-dinitrobenzaldehyde #, benzaldehyde,2-hydroxy-3,5-dinitro
Solubility Information Soluble in water,0.6g/100 mL (18°C).
Formula Weight 212.12
Percent Purity 98%
Chemical Name or Material 3,5-Dinitrosalicylaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 219-551-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only