missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3,5-Dinitrobenzyl Alcohol 98.0+%, TCI America™
$128.98 - $404.28
Chemical Identifiers
| CAS | 71022-43-0 |
|---|---|
| Molecular Formula | C7H6N2O5 |
| Molecular Weight (g/mol) | 198.13 |
| MDL Number | MFCD00007235 |
| InChI Key | GPHYIQCSMDYRGJ-UHFFFAOYSA-N |
| Synonym | 3,5-dinitrobenzyl alcohol, 3,5-dinitrophenyl methanol, benzenemethanol, 3,5-dinitro, 3,5-dinitrobenzenemethanol, unii-111z6y5ynw, 3,5-dinitrobenzylalcohol, pubchem20861, acmc-1bdpg, ksc494c8n, 3,5-dinitrophenyl methanol # |
| PubChem CID | 116978 |
| IUPAC Name | (3,5-dinitrophenyl)methanol |
| SMILES | OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O |
Chemical Identifiers
| 71022-43-0 | |
| 198.13 | |
| GPHYIQCSMDYRGJ-UHFFFAOYSA-N | |
| 116978 | |
| OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O |
| C7H6N2O5 | |
| MFCD00007235 | |
| 3,5-dinitrobenzyl alcohol, 3,5-dinitrophenyl methanol, benzenemethanol, 3,5-dinitro, 3,5-dinitrobenzenemethanol, unii-111z6y5ynw, 3,5-dinitrobenzylalcohol, pubchem20861, acmc-1bdpg, ksc494c8n, 3,5-dinitrophenyl methanol # | |
| (3,5-dinitrophenyl)methanol |
Specifications
| 71022-43-0 | |
| Yellow | |
| MFCD00007235 | |
| 3,5-dinitrobenzyl alcohol, 3,5-dinitrophenyl methanol, benzenemethanol, 3,5-dinitro, 3,5-dinitrobenzenemethanol, unii-111z6y5ynw, 3,5-dinitrobenzylalcohol, pubchem20861, acmc-1bdpg, ksc494c8n, 3,5-dinitrophenyl methanol # | |
| OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O | |
| 198.13 | |
| 198.13 | |
| Crystalline Powder |
| 91°C | |
| C7H6N2O5 | |
| 5 g | |
| GPHYIQCSMDYRGJ-UHFFFAOYSA-N | |
| (3,5-dinitrophenyl)methanol | |
| 116978 | |
| ≥98.0% (GC) | |
| 3,5-Dinitrobenzyl Alcohol |
Safety and Handling
TSCA : No