missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzyl alcohol, 98%

Catalog No. AAH3266922
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3266922 100 g
AAH3266906 5 g
AAH3266914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3266922 Supplier Thermo Scientific Chemicals Supplier No. H3266922
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 71022-43-0 | C7H6N2O5 | 198.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71022-43-0
Molecular Formula C7H6N2O5
Molecular Weight (g/mol) 198.13
MDL Number MFCD00007235
InChI Key GPHYIQCSMDYRGJ-UHFFFAOYSA-N
Synonym 3,5-dinitrobenzyl alcohol, 3,5-dinitrophenyl methanol, benzenemethanol, 3,5-dinitro, 3,5-dinitrobenzenemethanol, unii-111z6y5ynw, 3,5-dinitrobenzylalcohol, pubchem20861, acmc-1bdpg, ksc494c8n, 3,5-dinitrophenyl methanol #
PubChem CID 116978
IUPAC Name (3,5-dinitrophenyl)methanol
SMILES OCC1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 88°C to 91°C
Quantity 100 g
Beilstein 2054388
Formula Weight 198.13
Percent Purity 98%
Chemical Name or Material 3,5-Dinitrobenzyl alcohol
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 275-131-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only