missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dinitrobenzoyl Chloride 98.0+%, TCI America™

Catalog No. D082525G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D082525G 25 g
D0825500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D082525G Supplier TCI America Supplier No. D082525G

Chemical Identifiers

CAS 99-33-2
Molecular Formula C7H3ClN2O5
Molecular Weight (g/mol) 230.56
MDL Number MFCD00007248
InChI Key NNOHXABAQAGKRZ-UHFFFAOYSA-N
Synonym benzoyl chloride, 3,5-dinitro, 3,5-dinitrobenzoic acid chloride, dnbc, 3,5-dinitrobenzoylchloride, unii-5jfa2dvm4d, ccris 3137, 5jfa2dvm4d, 3,5-dinitrobenzoyl, acmc-209scd, dnbc;
PubChem CID 7432
IUPAC Name 3,5-dinitrobenzoyl chloride
SMILES C1=C(C=C(C=C1[N+](=O)[O-])[N+](=O)[O-])C(=O)Cl

Specifications

Melting Point 69°C
Color White-Yellow
Boiling Point 196°C
Quantity 25 g
UN Number 3261
Formula Weight 230.56
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3,5-Dinitrobenzoyl Chloride
RTECSNumber DM6637000
TSCA Yes