accessibility menu, dialog, popup

UserName

CAS RN 99-33-2

IUPAC Name: 3,5-dinitrobenzoyl chloride
Synonyms: benzoyl chloride, 3,5-dinitro-
Molecular Weight (g/mol): 230.56
Molecular Formula: C7H3ClN2O5
InChi Key: NNOHXABAQAGKRZ-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC(=C1)C(Cl)=O)[N+]([O-])=O
Boiling Point 
  • (2)
  • (2)
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
  • (1)
  • (3)
Quantity 
  • (2)
  • (3)
  • (1)
  • (2)
Physical Form 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (5)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)